Learn More
Invitrogen™ TIP60 Monoclonal Antibody (3F9)

Description
MA5-15346 targets TIP60 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15346 immunogen is purified recombinant fragment of human TIP60 expressed in E. Coli. MA5-15346 detects TIP60 which has a predicted molecular weight of approximately 60kDa.
Specifications
Specifications
| Antigen | TIP60 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 3F9 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.03% sodium azide |
| Gene | KAT5 |
| Gene Accession No. | Q92993 |
| Gene Alias | 60 kDa Tat-interactive protein; AI839539; cPLA(2)-interacting protein; CPLA2; cPLA2 interacting protein; ESA1; histone acetyltransferase HTATIP; Histone acetyltransferase KAT5; HIV-1 Tat interactive protein; HIV-1 tat interactive protein 1, 60 kDa homolog; HIV-1 tat interactive protein 1, homolog; HIV-1 Tat interactive protein 60 kD; HIV-1 Tat interactive protein, 60 kD; HIV-1 Tat interactive protein, 60kDa; HIV-1 tat interactive protein, homolog; HTATIP; Htatip1; K(lysine) acetyltransferase 5; K-acetyltransferase 5; Kat5; Lysine acetyltransferase 5; PLIP; Tat interacting protein, 60kDa; Tat-interactive protein-60; TIP; Tip55; Tip60; TIP60(HTATIP); Tip60b; ZC2HC5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.