Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIPARP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116171
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116171 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116171 Supplier Invitrogen™ Supplier No. PA5116171
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TIPARP Polyclonal Antibody for Western Blot

TIPARP is a poly [ADP-ribose] polymerase using NAD+ as a substrate to transfer ADP-ribose onto glutamic acid residues of a protein acceptor; repeated rounds of ADP-ribosylation leads to the formation of poly(ADPribose) chains on the protein, thereby altering the function of the target protein. TIPARP may play a role in the adaptative response to chemical exposure (TCDD) and thereby mediates certain effects of the chemicals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIPARP
Applications Western Blot
Classification Polyclonal
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TIPARP
Gene Accession No. Q7Z3E1, Q8C1B2
Gene Alias ADP-ribosyltransferase diphtheria toxin-like 14; ARTD14; AW558171; DDF1; DKFZP434J214; DKFZp686N0351; DKFZp686P1838; FLJ40466; PARP-1; PARP7; PARP-7; pART14; Poly [ADP-ribose] polymerase 7; protein mono-ADP-ribosyltransferase TIPARP; RM1; TCDD inducible poly(ADP-ribose) polymerase; TCDD-inducible poly [ADP-ribose] polymerase; TCDD-inducible poly(ADP-ribose) polymerase; TIPARP
Gene Symbols TIPARP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 263-292 amino acids from the C-terminal region of human Tiparp.
Purification Method Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25976, 99929
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.