Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIPIN Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298947G
Description
TIPIN Polyclonal specifically detects TIPIN in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
TIPIN | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
DyLight 488 | |
FLJ20516, TIMELESS interacting protein, TIMELESS-interacting protein | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human TIPIN. | |
0.1 mL | |
DNA Repair | |
54962 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction