Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIPIN Monoclonal Antibody (OTI3A1)
GREENER_CHOICE

Catalog No. PIMA526569
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526569 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526569 Supplier Invitrogen™ Supplier No. MA526569
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIPIN Monoclonal Antibody for Western Blot, IHC (P)

Required for normal progression of S-phase. Important for cell survival after DNA damage or replication stress. May be specifically required for the ATR-CHK1 pathway in the replication checkpoint induced by hydroxyurea or ultraviolet light.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIPIN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3A1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TIPIN
Gene Accession No. Q9BVW5
Gene Alias 1110005A05Rik; 1110018P21Rik; CSM3 homolog; FLJ20516; RGD1564084; TIMELESS interacting protein; TIMELESS-interacting protein; TIMELESS-interacting-like protein; tipin; Unknown (protein for MGC:127806); zgc:91928
Gene Symbols TIPIN
Host Species Mouse
Immunogen Full length human recombinant protein of TIPIN produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54962
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.