Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIPIN Polyclonal Antibody

TIPIN Polyclonal Antibody
Supplier: Thermo Scientific PA5112079
Description
TIPIN Polyclonal Antibody for Western Blot, ICC/IF, IP
Specifications
| TIPIN | |
| Polyclonal | |
| Unconjugated | |
| TIPIN | |
| 1110005A05Rik; 1110018P21Rik; CSM3 homolog; FLJ20516; RGD1564084; TIMELESS interacting protein; TIMELESS-interacting protein; TIMELESS-interacting-like protein; tipin; Unknown (protein for MGC:127806); zgc:91928 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 54962 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9BVW5 | |
| TIPIN | |
| A synthetic peptide corresponding to the C-terminus of the Human TIPIN. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction