Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIPIN Polyclonal Antibody

Catalog No. PIPA535995
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535995 Supplier Invitrogen™ Supplier No. PA535995
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2, human TIPIN-transfected 293T cells. Predicted reactivity: Mouse (83%), Rat (84%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Required for normal progression of S-phase. Important for cell survival after DNA damage or replication stress. May be specifically required for the ATR-CHK1 pathway in the replication checkpoint induced by hydroxyurea or ultraviolet light.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIPIN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene TIPIN
Gene Accession No. Q9BVW5
Gene Alias 1110005A05Rik; 1110018P21Rik; CSM3 homolog; FLJ20516; RGD1564084; TIMELESS interacting protein; TIMELESS-interacting protein; TIMELESS-interacting-like protein; tipin; Unknown (protein for MGC:127806); zgc:91928
Gene Symbols TIPIN
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TIPIN. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54962
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.