Learn More
TIPIN Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301474A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Required for normal progression of S-phase. Important for cell survival after DNA damage or replication stress. May be specifically required for the ATR-CHK1 pathway in the replication checkpoint induced by hydroxyurea or ultraviolet light.Specifications
TIPIN | |
Polyclonal | |
Unconjugated | |
TIPIN | |
TIMELESS interacting protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54962 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9BVW5 | |
TIPIN | |
Between 251 and 301 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.