Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tiplaxtinin
GREENER_CHOICESDP

Catalog No. 556550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
556550 50 mg
556510 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 556550 Supplier Tocris Bioscience™ Supplier No. 5565/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Plasminogen activator inhibitor-1 (PAI-1) inhibitor

Tiplaxtinin is a plasminogen activator inhibitor-1 (PAI-1) inhibitor (IC50 = 2.7 μM). Induces vascular smooth muscle cell apoptosis in vitro and reduces carotid artery neointimal formation in a rat vascular injury model. Promotes thrombolysis in rat and dog models of acute arterial thrombosis. Orally available.

Specifications

Chemical Name or Material α-Oxo-1-(phenylmethyl)-5-[4-(trifluoromethoxy)phenyl]-1H-indole-3-acetic acid
CAS 393105-53-8
Synonym PAI 039, tiplasinin
Quantity 50 mg
Target Additional Synthetic Protease Inhibitors
Molecular Formula C24H16F3NO4
Formula Weight 439.38
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.