Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIPRL Antibody (OTI3D11), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274529PE
Description
TIPRL Monoclonal specifically detects TIPRL in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
TIPRL | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
TIPRL | |
Full length recombinant protein of human TIPRL (NP_690866) produced in HEK293T cells. | |
0.1 mL | |
261726.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI3D11 | |
Flow Cytometry | |
dJ69E11.3, MGC3794, putative MAPK activating protein, Putative MAPK-activating protein PM10, TIP41, TOR signaling pathway regulator-like (S. cerevisiae), TIP41-like protein, TIP41Type 2A-interacting protein, TIPTIP41, TOR signalling pathway regulator-like (S. cerevisiae) | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction