Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIPRL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555746
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555746 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555746 Supplier Invitrogen™ Supplier No. PA555746
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SHRDFCFGPW KLTASKTHIM KSADVEKLAD ELHMPSLPEM MFGDNVLRIQ HGSGFGIEFN ATDALRCVNN YQGMLK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

TIPRL is an inhibitory regulator of protein phosphatase-2A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIPRL
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TIPRL
Gene Accession No. O75663
Gene Alias 1810011K17Rik; CG9578-like; dJ69E11.3; MGC3794; OTTHUMP00000032581; OTTHUMP00000032582; putative MAPK-activating protein PM10; RGD1310442; TIP; TIP41; TIP41, TOR signaling pathway regulator-like; TIP41, TOR signalling pathway regulator-like (S. cerevisiae); TIP41-like protein; Tiprl; TIPRL1; TOR signaling pathway regulator; type 2A-interacting protein
Gene Symbols TIPRL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TIPRL. Recombinant protein control fragment (Product #RP-94269).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 261726
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.