Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIPRL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519241
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519241 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519241 Supplier Invitrogen™ Supplier No. PA519241
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

TIPRL is an inhibitory regulator of protein phosphatase-2A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIPRL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene TIPRL
Gene Accession No. O75663
Gene Alias 1810011K17Rik; CG9578-like; dJ69E11.3; MGC3794; OTTHUMP00000032581; OTTHUMP00000032582; putative MAPK-activating protein PM10; RGD1310442; TIP; TIP41; TIP41, TOR signaling pathway regulator-like; TIP41, TOR signalling pathway regulator-like (S. cerevisiae); TIP41-like protein; Tiprl; TIPRL1; TOR signaling pathway regulator; type 2A-interacting protein
Gene Symbols TIPRL
Host Species Goat
Immunogen Synthetic peptide sequence (RIDPNPADSQKSTQ) corresponding to the C-terminus amino acids of TIPRL (aa 257-271).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 261726
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.