Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIPRL Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501979955
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9955 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9955 Supplier Bioss Supplier No. BS19121R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TIPRL is an inhibitory regulator of protein phosphatase-2A .

Specifications

Antigen TIPRL
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 15mM sodium azide; pH
Gene TIPRL
Gene Accession No. O75663
Gene Alias 1810011K17Rik; CG9578-like; dJ69E11.3; MGC3794; OTTHUMP00000032581; OTTHUMP00000032582; Putative MAPK-activating protein PM10; TIP; TIP41; TIP41, TOR signaling pathway regulator-like; TIP41, TOR signalling pathway regulator-like (S. cerevisiae); TIP41-like protein; TIPRL; TIPRL1; TOR signaling pathway regulator; Type 2A-interacting protein
Gene Symbols TIPRL
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TIPRL.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 261726
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.