Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VistaLab™ Technologies Pipette Tips: Low Retention

Shop All VistaLab Technologies Inc. Products


Click to view available options
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Volume (Metric):
2 μL
25 μL
250 μL
1250 μL
Description
- Sterile, Low Retention tips packaged in VistaRak: 5 racks of 192 tips each
- Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, etc.
- Use VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation pipettes
- All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time
Specifications
Specifications
Description | Low Retention |
Volume (Metric) | 1250 μL |
Filtered | Non-filtered |
Sterility | Non-sterile |
Color | Clear |
Rack Color | Blue |
Autoclavable | Autoclavable |
Length (English) | 2.2 in. |
Thickness | 5 in. |
For Use With (Application) | Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, TRIS, BSA and Tween, PCR or other biological applications |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction