Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies Pipette Tips: Low Retention
GREENER_CHOICESDP

Catalog No. 22147631 Shop All VistaLab Technologies Inc. Products
Click to view available options
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Volume (Metric):
2 μL
25 μL
250 μL
1250 μL

For use with Ovation and MLA Brand pipetters

  • Sterile, Low Retention tips packaged in VistaRak: 5 racks of 192 tips each
  • Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, etc.
  • Use VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation pipettes
  • All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time

Specifications

Description Low Retention
Volume (Metric) 1250 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Rack Color Blue
Autoclavable Autoclavable
Length (English) 2.2 in.
Thickness 5 in.
Certifications/Compliance Certified RNase/DNase free, non-pyrogenic
For Use With (Application) Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, TRIS, BSA and Tween, PCR or other biological applications
Sterilization Method e-beam
Compatibility MLA/Ovation Pipettes
Format Rack
Tip Style Low Retention, Straight
Packaging VistaRak: 5 racks
Quantity 192 Tips/Rack
Product Type Pipette Tips
Color Code Blue
Disposable Single-use
For Use With (Equipment) MLA, Ovation
Graduated No
Material Polypropylene
Shape Cone
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.