Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIRAP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | TIRAP |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
TIRAP Polyclonal specifically detects TIRAP in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TIRAP | |
Polyclonal | |
Rabbit | |
Human | |
adapter protein wyatt, Adaptor protein Wyatt, MAL, MyD88 adapter-like protein, MyD88-2, TIR domain-containing adapter protein, toll/interleukin-1 receptor domain-containing adapter protein, toll-interleukin 1 receptor (TIR) domain containing adaptor protein, Toll-interleukin 1 receptor (TIR) domain-containing adaptor protein, Toll-like receptor adaptor protein, wyatt | |
TIRAP | |
IgG | |
Affinity Purified |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
114609 | |
This antibody was developed against a recombinant protein corresponding to amino acids: SSRWSKDYDVCVCHSEEDLVAAQDLVSYLEGSTASLRCFLQLRDATPGGAIVSELCQALSSSHCRVLLITPGFLQDPWCKYQML | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title