Learn More
Novus Biologicals™ TIRAP (TLR2 and TLR4) Inhibitor Peptide Set

Description
TLR2 / TLR4 Inhibitor Peptide: 1mg (lyophilized); sequence: DRQIKIWFQNRRMKWKKPGFLRDPWCKYQML (Inhibitor sequence: PGFLRDPWCKYQML); Molecular weight: 4097.92 Da ; Antennapedia Control Peptide: 1mg (lyophilized); sequence: DRQIKIWFQNRRMKWKK; Molecular weight: 2361 Da
Specifications
Specifications
| Color | White |
| Components | TLR2 / TLR4 Inhibitor Peptide: 1mg (lyophilized); sequence: DRQIKIWFQNRRMKWKKPGFLRDPWCKYQML (Inhibitor sequence: PGFLRDPWCKYQML); Molecular weight: 4097.92 Da ; Antennapedia Control Peptide: 1mg (lyophilized); sequence: DRQIKIWFQNRRMKWKK; Molecular weight: 2361 Da |
| For Use With (Application) | Inhibit TLR2 and TLR4 signaling |
| Content And Storage | Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
| Quantity | 1 mg |
| Product Type | TIRAP (TLR2 and TLR4) Inhibitor Peptide Set |
| Inhibitors | TLR2 and TLR4 |
| Form | Solid |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.