Learn More
Specifications
Specifications
| Host Species | Human, Mouse |
| Components | TIRAP Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKLQLRDAAPGGAIVS (TIRAP sequence is underlined). Molecular weight: 3701.4. Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361. |
| For Use With (Application) | Inhibition of TIRAP binding to TLR2 or TLR4 |
| Content And Storage | Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
| Quantity | 5 mg |
| Product Type | TIRAP (TLR2 and TLR4) Inhibitor Peptide Set |
| Molecular Weight (g/mol) | 3701.4 |
| Product Line | Novus |
| Inhibitors | TLR2, TLR4 |
| Form | Lyophilized |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
