Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ringers Tire Buddy Gloves

Catalog No. 19050922B Shop All Ansell Products
Encompass
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-050-922B Medium
19-050-922C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-050-922B Supplier Ringers Supplier No. 14309
This item is not returnable. View return policy

Reduce wear on the hands with a high-abrasion resistant palm made from EVA foam padding to minimize fatigue.

  • High-abrasion resistant palm reduces wear on the hands
  • EVA foam padding in lower portion of palm to reduce fatigue and injuries
  • Tough spandex top material
  • Rubberized Clarino™ Zeus fingers
  • Open elastic cuff allows for easy access
TRUSTED_SUSTAINABILITY

Specifications

Back Material Spandex
Cuff Material Elastic
Coating Material Uncoated
Color Black
Hand Specific
Material Synthetic Leather
Product Type Mechanic's Gloves
Size Medium
For Use With (Application) Automotive Assembly
Glove Cuff Style Open
Series Tire Buddy
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.