Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tiron, Indicator, Solstice

Catalog No. 60047723 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-723 10 g Plastic bottle
60-047-722 100 g Plastic bottle
60-047-724 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-047-723 Supplier Solstice Supplier No. 8946010G

Indicator (for complexometry)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 270573-71-2
Molecular Formula C6H6Na2O9S2
Molecular Weight (g/mol) 332.205
MDL Number MFCD00149531
InChI Key ZLRROLLKQDRDPI-UHFFFAOYSA-L
PubChem CID 2723960
IUPAC Name disodium;4,5-dihydroxybenzene-1,3-disulfonate;hydrate
SMILES C1=C(C=C(C(=C1S(=O)(=O)[O-])O)O)S(=O)(=O)[O-].O.[Na+].[Na+]

Specifications

Melting Point 300°C
Linear Formula (OH)2C6H2(SO3Na)2 · H2O
UN Number NONH for all modes of transport
Solubility Information 1350g/L
Formula Weight 332.22g/mol
Packaging Plastic bottle
Color Violet to White
pH 5.0
Quantity 10 g
Physical Form Solid
Chemical Name or Material Tiron
Show More Show Less
Hazard Statement P261-P305 + P351 + P338
RTECSNumber 205-741-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.