Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIS11D Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297954MFV610
Description
TIS11D Polyclonal antibody specifically detects TIS11D in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
TIS11D | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
HF.10, HF10Zinc finger protein HF.10, Zfp105, zinc finger protein 35, zinc finger protein 35 (clone HF.10) | |
Produced in rabbits immunized with E. coli-derived Human TIS11D fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
678 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction