Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIS11D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538612
Description
TIS11D, also known as ZFP36LA (zinc finger protein 36, C3H type-like 2), BRF2 (butyrate response factor 2), ERF2 or RNF162C, is a 494 amino acid protein that localizes to the nucleus and contains two CSH1-type zinc fingers. Belonging to the TIS11 family of early response proteins, TIS11D is thought to function as a nuclear transcription factor that binds to 5′UUAUUUAUUU-3′ core RNA sequences and may regulate growth factor-induced cellular responses. The gene encoding TIS11D maps to human chromosome 2, which houses over 1,400 genes and comprises nearly 8% of the human genome. Harlequin icthyosis, a rare and morbid skin deformity, is associated with mutations in the ABCA12 gene, while the lipid metabolic disorder sitosterolemia is associated with defects in the ABCG5 and ABCG8 genes.
Specifications
| TIS11D | |
| Polyclonal | |
| Unconjugated | |
| ZFP36L2 | |
| TISD, Zinc finger protein 36, C3H1 type-like 2, ZFP36-like 2, Butyrate response factor 2, EGF-response factor 2, ERF-2, Protein TIS11D, ZFP36L2, BRF2, ERF2, RNF162C, TIS11D | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12193, 678 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P47974, P23949 | |
| ZFP36L2 | |
| A synthetic peptide derived from the internal region of human TISD | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction