Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIS11D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101115
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101115 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101115 Supplier Invitrogen™ Supplier No. PA5101115
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TISD.

TIS11D, also known as ZFP36LA (zinc finger protein 36, C3H type-like 2), BRF2 (butyrate response factor 2), ERF2 or RNF162C, is a 494 amino acid protein that localizes to the nucleus and contains two CSH1-type zinc fingers. Belonging to the TIS11 family of early response proteins, TIS11D is thought to function as a nuclear transcription factor that binds to 5'UUAUUUAUUU-3' core RNA sequences and may regulate growth factor-induced cellular responses. The gene encoding TIS11D maps to human chromosome 2, which houses over 1, 400 genes and comprises nearly 8% of the human genome. Harlequin icthyosis, a rare and morbid skin deformity, is associated with mutations in the ABCA12 gene, while the lipid metabolic disorder sitosterolemia is associated with defects in the ABCG5 and ABCG8 genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIS11D
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZFP36L2
Gene Accession No. P23949, P47974
Gene Alias Brf2; Butyrate response factor 2; EGF-response factor 2; ERF2; ERF-2; LOW QUALITY PROTEIN: mRNA decay activator protein ZFP36L2; mRNA decay activator protein ZFP36L2; Protein TIS11D; RNF162C; TIS11D; TISD; TPA-induced sequence 11d; ZFP36 ring finger protein like 2; ZFP36 ring finger protein-like 2; ZFP36L2; ZFP36-like 2; zinc finger protein 36, C3H type-like 1; zinc finger protein 36, C3H type-like 2; zinc finger protein 36, C3H type-like 2-like; zinc finger protein 36, C3H1 type-like 2; zinc finger protein, C3H type, 36-like 2
Gene Symbols ZFP36L2
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZFP36L2(Accession P47974), corresponding to amino acid residues T238-P288.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12193, 678
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.