Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tissue alpha-L-Fucosidase/FUCA1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP321436C
Description
Tissue alpha-L-Fucosidase/FUCA1 Polyclonal antibody specifically detects Tissue alpha-L-Fucosidase/FUCA1 in Human samples. It is validated for ELISASpecifications
Tissue alpha-L-Fucosidase/FUCA1 | |
Polyclonal | |
ELISA | |
Alpha-L-fucosidase 1, Alpha-L-fucosidase I, Alpha-L-fucoside fucohydrolase 1, EC 3.2.1, EC 3.2.1.51, FUCA, fucosidase, alpha-L- 1, tissue, tissue alpha-L-fucosidase | |
Produced in rabbits immunized with purified, recombinant Human Tissue alpha-L-Fucosidase/FUCA1 (Gln32-Lys466; Uniprot: P04066) | |
0.1 mL | |
Signal Transduction | |
2517 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction