Learn More
Tocris Bioscience™ Tissue Clearing Pro-Organoid
Description
A wide range of IHC validated antibodies are available from R&D Systems and Novus Biologicals ; optimization of antibody incubation conditions for use with the Tissue Clearing Pro-Organoid technique is recommended.
Please read the Tissue Clearing Pro-Organoid guidebook for more information and detailed protocols for using this kit.
Specifications
Specifications
| Quantity | 1 Pk. |
| Target | General Laboratory Research Reagent |