Learn More
R&D Systems™ Tissue Clearing Pro Reagent 1
SDP
Catalog No. 75631PACK Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Pk.
Catalog No. Quantity
75631PACK 1 Pk.
1 options
This item is not returnable. View return policy
Catalog No. 75631PACK Supplier R&D Systems™ Supplier No. 7563/1 PACK
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
A Tocris product, part of R&D Systems. Tissue clearing reagent
Tissue Clearing Pro is a solvent-based tissue clearing technique, comparable to BABB and DISCO. It does not cause significant shrinkage or tissue damage and does not use as harsh a solvent as other solvent-based techniques. Tissue Clearing Pro Reagent 1 is part of our Tissue Clearing Pro kit (Cat. No. 7389) that can clear a whole mouse brain of up to 8 mm thickness in 24 hours; for sections with a thickness of 1 mm this can be achieved in 2 hours. For small tissues and thin brain slices (500 μm) that do not require permeabilization, only Tissue Clearing Pro Reagent 1 is required and tissues can be imaged in Tissue Clearing Pro Reagent 1. Larger tissues must be placed in Tissue Clearing Pro Reagent 2 (Cat. No. 7564) to finish clearing, then these samples can be mounted and imaged in Tissue Clearing Pro Reagent 2. This product is compatible with fluorescent protein, immunolabeling and small molecule staining techniques, as well as being suitable for use with all standard dyes and buffers. Following 3D imaging, clearing can be reversed for follow-up 2D histology staining. Tissue Clearing Pro Reagent 1 has a refractive index of 1.50.
Please read the Tissue Clearing Pro Guidebook for more information.
Specifications
Quantity 1 Pk.
Product Line Tocris
Target General Laboratory Research Reagent