Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tissue Clearing Pro Reagent 2

Catalog No. 75641PACK
Change view
Click to view available options
Quantity:
1 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75641PACK 1 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 75641PACK Supplier Tocris Bioscience™ Supplier No. 7564/1 PACK
This item is not returnable. View return policy

Tissue clearing reagent

Tissue Clearing Pro is a solvent-based tissue clearing technique, comparable to BABB and DISCO. It does not cause significant shrinkage or tissue damage and does not use as harsh a solvent as other solvent-based techniques. Tissue Clearing Pro Reagent 2 is part of our Tissue Clearing Pro kit (Cat. No. 7389) that can clear a whole mouse brain of up to 8 mm thickness in 24 hours; for sections with a thickness of 1 mm this can be achieved in 2 hours. For small tissues and thin brain slices (500 μm) that do not require permeabilization, only Tissue Clearing Pro Reagent 1 (Cat. No. 7563) is required and tissues can be imaged in Tissue Clearing Pro Reagent 1. Larger tissues must be placed in Tissue Clearing Pro Reagent 2 to finish clearing, then these samples can be mounted and imaged in Tissue Clearing Pro Reagent 2. This product is compatible with fluorescent protein, immunolabeling and small molecule staining techniques, as well as being suitable for use with all standard dyes and buffers. Following 3D imaging, clearing can be reversed for follow-up 2D histology staining. Tissue Clearing Pro Reagent 2 has a refractive index of 1.53.

Please read the Tissue Clearing Pro Guidebook for more information.

Specifications

Quantity 1 Pk.
Target General Laboratory Research Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.