Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tissue Clearing Pro

Catalog No. 73891PACK
Change view
Click to view available options
Quantity:
1 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73891PACK 1 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 73891PACK Supplier Tocris Bioscience™ Supplier No. 7389/1 PACK
This item is not returnable. View return policy

Tissue clearing reagent kit

Tissue Clearing Pro is a rapid, easy-to-use, reversible and non-destructive clearing and staining kit, which allows effective tissue clearing and staining of whole tissues. This kit can clear a whole mouse brain of up to 8 mm thickness in 24 hours; for sections with a thickness of 1 mm this can be achieved in 2 hours. Tissue Clearing Pro is compatible with fluorescent protein, immunolabeling and small molecule staining techniques, as well as being suitable for use with all standard dyes and buffers. Following 3D imaging, Tissue Clearing Pro can be reversed for follow-up 2D histology staining. The kit comes with a getting started guide and a more comprehensive tissue clearing guidebook, providing detailed protocols. The Tissue Clearing Pro kit contains 30 mL of Tissue Clearing Pro clearing reagent 1 and 2, plus the 5 buffer solutions required to process tissues for immunolabeling, and is enough to process four whole mouse brains (approx. 2 g of tissue).

A wide range of IHC validated antibodies are available from R&D Systems and Novus Biologicals ; optimization of antibody incubation conditions for use with the Tissue Clearing Pro technique is recommended.

Please read the Tissue Clearing Pro Guidebook for more information and detailed protocols for using this kit.

Specifications

Quantity 1 Pk.
Target General Laboratory Research Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.