Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCI America™ Tissue-Clearing Reagent CUBIC-B
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Capacity:
25 mL
100 mL
Description
- The CUBIC technique enables cyclopedic imaging at a single-cell resolution following whole-body and whole-organ clearing.
- CUBIC-B is an optional addition to the CUBIC clearing protocol, and can be used to clear bone tissue.
Note: Both T3741 and T3740 are required for the CUBIC clearing process.
Specifications
Specifications
Capacity | 100 mL |
For Use With (Equipment) | Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device |
Product Line | CUBIC |
For Use With (Application) | Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction