Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCI America™ Tissue-Clearing Reagent CUBIC-HL
SDP

Catalog No. p-7222736 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Capacity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
T378125ML 25 mL
T3781100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T378125ML Supplier TCI America™ Supplier No. T378125ML
Only null left
Add to Cart
Add to Cart

CUBIC-HL is optimized for clearing highly fatty tissues. Note this does not preserve the fluorescent signal of fluorescent proteins.

  • The CUBIC technique enables cyclopedic imaging at a single-cell resolution following whole-body and whole-organ clearing.
  • CUBIC-HL is an optional addition to the CUBIC clearing protocol, and can be used to clear highly fatty tissue and quenching autofluourescence.

Note: Both T3741 and T3740 are required for the CUBIC clearing process.

Specifications

Capacity 25 mL
For Use With (Equipment) Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device
Product Line CUBIC
For Use With (Application) Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.