Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCI America™ Tissue-Clearing Reagent CUBIC-P
SDP

Catalog No. T3782100ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Capacity:
25 mL
100 mL

CUBIC-P is a CUBIC reagent for mouse perfusion, which efficiently aids perfusion fixation.

  • The CUBIC technique enables cyclopedic imaging at a single-cell resolution following whole-body and whole-organ clearing.
  • CUBIC-P is an optional addition to the CUBIC clearing protocol, and can be used to improve perfusion fixation.

Note: Both T3741 and T3740 are required for the CUBIC clearing process.

Specifications

Capacity 100 mL
For Use With (Equipment) Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device
Product Line CUBIC
For Use With (Application) Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.