Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCI America™ Tissue-Clearing Reagent iTOMEI-D
SDP

Catalog No. p-7222743 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Capacity:
5 mL
25 mL

Catalog No. T39405ML

Supplier: TCI America™ T39405ML

Only null left
Add to Cart
Add to Cart

Improved TOMEI (iTOMEI) is a method that specializes the TOMEI clearing process for fluorescent protein observation.

  • Enables transparency by simple osmosis in just a few days
  • Keeps the fluorescence of a fluorescent protein such as GFP or tdTomato co-expressed as a reporter gene
  • Suppresses auto-fluorescence
  • Applicable to wide variety of plant species including Oryza sativa, Arabidopsis thaliana, Marchantia polymorpha

Specifications

Capacity 5 mL
For Use With (Equipment) Confocal Laser-scanning Microscopy (CLSM) Device
Product Line TOMEI
For Use With (Application) Plant Cytology and Histological research
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.