Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Tissue Culture Rotator

Catalog No. p-3606721 Shop All Fisherbrand Products
Click to view available options
Holds:
142 Tubes
Tube Size:
17 mm dia.

Catalog No. 14-251-251

Only null left
Add to Cart
Add to Cart

Requires

Rotator Body (order separately)

Single-drum unit gently rotates up to 142 samples.

Designed for growing tissue and virus cultures, hormone production, sample testing for the presence of viruses, and cytotoxicity assays.

  • Variable rotation from 5 to 70rpm
  • Elevation angle is adjustable from 5° above horizontal to 90°
  • Drum can be lifted off roller without interrupting operation
  • POWER-ON pilot light
  • Can be used in standard incubators under controlled temperature conditions up to 40°C and humidity conditions from 20 to 80% noncondensing
  • Powder-coated stainless-steel outer body is rugged and corrosion-resistant
  • Long-life maintenance-free, brushless motor

Order Info

For a complete system, you must order rotator body (14-251-250) and one drum. Note: the Rotator Body (14-251-250) is currently discontinued and is no longer available. Dimensions (rotator body): 15D x 14W x 22.5 in. H (38 x 36 x 57cm). Unit weight (without roller): 19 lb. (9kg). All drums are 14 in. dia. (36cm) and weigh 5 lb. (2kg).

Warranty and Services

24 months

TRUSTED_SUSTAINABILITY

Specifications

Holds 142 Tubes
Tube Size 17 mm dia.
Product Type Rotator Drum
Diameter (Metric) 36 cm
Length (English) 14.17 in.
Length (Metric) 36 cm
For Use With (Application) Designed for growing tissue and virus cultures, hormone production, sample testing for the presence of viruses and cytotoxicity assays
No. of Positions 142
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.