Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tissue Dry Dehydrant, Fisher Chemical

Catalog No. sh54 Shop All Fisherbrand Products
Click to view available options
Quantity:
4 L
This item is not returnable. View return policy

Catalog No. SH5-4

Only null left
Add to Cart
This item is not returnable. View return policy

Specifications

Physical Form Liquid
Chemical Name or Material Tissue Dry Dehydrant
CAS 109-86-4
Packaging Aluminum Can
Color Colorless
Quantity 4 L
MDL Number MFCD00212754
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.