Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Tissue Forceps
SDP

Tissue forceps with teeth for holding tissue during surgery.

$73.10 - $125.30

Specifications

For Use With (Application) Neuroscience
View More Specs

Products 14
Catalog Number Mfr. No. Length (English) Model Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Model Price Quantity & Availability  
10-001-274
SDP
View Documents
Stoelting™
5210867
4.5 in. 1 x 2 teeth, Delicate
Each for $97.10
Only null left
Add to Cart
 
10-001-278
SDP
View Documents
Stoelting™
5210871
4.5 in. 2 x 3 teeth
Each for $105.90
Only null left
Add to Cart
 
10-000-458
SDP
View Documents
Stoelting™
5210860P
4.5 in. 1 x 2 teeth
Each for $74.60
Only null left
Add to Cart
 
10-001-275
SDP
View Documents
Stoelting™
5210868
5 in. 1 x 2 teeth, Delicate
Each for $103.30
Only null left
Add to Cart
 
10-001-279
SDP
View Documents
Stoelting™
5210872
5 in. 2 x 3 teeth
Each for $103.30
Only null left
Add to Cart
 
10-001-280
SDP
View Documents
Stoelting™
5210873
5.5 in. 2 x 3 teeth
Each for $105.90
Only null left
Add to Cart
 
10-001-272
SDP
View Documents
Stoelting™
5210864
5.5 in. 1 x 2 teeth
Each for $95.70
Only null left
Add to Cart
 
10-000-460
SDP
View Documents
Stoelting™
5210869P
5.75 in. 1 x 2 teeth
Each for $91.70
Only null left
Add to Cart
 
10-000-459
SDP
View Documents
Stoelting™
5210862P
5 in. 1 x 2 teeth
Each for $74.60
Only null left
Add to Cart
 
10-000-461
SDP
View Documents
Stoelting™
5210880P
5 in. 3 x 4 teeth
Each for $73.10
Only null left
Add to Cart
 
10-001-277
SDP
View Documents
Stoelting™
5210870
6 in. 1 x 2 teeth, Delicate
Each for $113.30
Only null left
Add to Cart
 
10-001-281
SDP
View Documents
Stoelting™
5210874
6 in. 2 x 3 teeth
Each for $103.30
Only null left
Add to Cart
 
10-001-273
SDP
View Documents
Stoelting™
5210866
6 in. 1 x 2 teeth
Each for $95.70
Only null left
Add to Cart
 
10-001-285
SDP
View Documents
Stoelting™
5210884
6 in. 3 x 4 teeth
Each for $125.30
Only null left
Add to Cart
 
Description

Description

Specifications

Specifications

Neuroscience
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.