Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
StatLab™ Tissue Margin Marking Dyes



Click to view available options
Color:
Black
Blue
Green
Orange
Red
Red, Orange, Yellow, Green, Blue, Violet, Black
Violet
Yellow
Format:
Bottle
Set
Description
- Durable, bright dyes hold permanently through processing and staining procedures
- Unique lids may be completely removed or conveniently flipped open to prevent drying
- Available in 7 dye colors
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Capacity | 2 oz. |
Color | Orange |
For Use With (Application) | Defining Tissue Margins |
Format | Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction