Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
StatLab™ Tissue Marking Dye

Click to view available options
Color:
Black
Blue
Green
Orange
Red
Violet
Yellow
Description
- Proprietary formulation designed to eliminate evaporation and sludgy dyes, with fewer drips and spills
- Durable, bright dyes hold permanently through processing and staining procedures
- Unique lids may either be completely removed or conveniently flipped open to prevent drying
Specifications
Specifications
Color | Red |
Product Type | Tissue Marking Dye |
Quantity | 8 oz. Bottle |
Solution Type | Mordant Dye |
For Use With (Application) | Tissue Marking |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction