Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Tissue Path™ IV Lids and Bases

Catalog No. 22047065 Shop All Fisherbrand Products
Click to view available options
Color:
Aqua
Lilac
Peach
Pink
White
Yellow
Product Type:
Embedding Cassette Base
Tissue Cassette Base
Tissue Cassette Lid
This item is not returnable. View return policy
This item is not returnable. View return policy

For SHUR/Mark™ and other automatic labeling systems

  • Lids are available 1000 per box, two boxes per case
  • Bases are available 500 per box, four boxes per case

Specifications

Color Aqua
Material Acetyl Polymer
Product Type Tissue Cassette Lid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.