Learn More
Thermo Scientific™ Tissue Protein Extraction Bundle PROMO
Click to view available options
:
Thermo Scientific™ Tissue Protein Extraction Bundle
Description
- T-PER tissue cell lysis reagent utilizes a mild detergent in 25mM bicine, 150mM sodium chloride (pH 7.6) to maximize the efficiency of protein solubilization from mammalian tissue samples by homogenization. Cell lysates prepared with T-PER Reagent are directly compatible with reporter assays (e.g., luciferase, beta-galactosidase, chloramphenicol acetyl transferase), protein kinase assays (e.g., PKA, PKC, tyrosine kinase), immunoassays (e.g., Western blots, ELISAs, RIAs) and/or protein purification procedures
- Pierce Protease Inhibitor Mini Tablets protects proteins from proteolytic degradation through inhibition of serine proteases, cysteine proteases, aspartic acid proteases, and aminopeptidases that are typically present in cell lysate samples
- Pierce Rapid Gold BCA Protein Assay Kit is a two-component, high-precision, detergent-compatible assay optimized to measure (at 480 nm) total protein concentration compared to a protein standard curve of known concentrations. The assay uses the same copper-chelating technology as the well-known traditional Pierce BCA Protein Assay and provides comparable accuracy but with a 5 minute, room temperature incubation. This improvement eliminates the need to wait or expose the samples to elevated temperatures for a fast time to results
Specifications
Specifications
Quantity | 1 Bundle |
Includes | T-PER Tissue Protein Extraction Reagent (500 mL), Pierce Protease Inhibitor Mini Tablets (30 tablets), Pierce Rapid Gold BCA Protein Assay Kit (250 mL) |
Content And Storage | See instructions for individual components |