Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Tissue Slicer
SDP

Stoelting™ Tissue Slicer is designed to prepare thin slices of living brain and is also excellent for slicing other tissues, blocking fixed tissue for histology, and slicing electrophoretic gels.

$1715.00 - $2960.00

Specifications

For Use With (Application) Neuroscience
View More Specs

Products 2
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
10-000-126
SDP
View Documents
Stoelting™
51415
Tissue slicer with digital
Each for $2,960.00
Only null left
Add to Cart
 
10-000-127
SDP
View Documents
Stoelting™
51425
Tissue slicer
Each for $1,715.00
Only null left
Add to Cart
 
Description

Description

Sections as thin as 150 microns can be reliably prepared.
Specifications

Specifications

Neuroscience
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.