Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TissuePrep™ 2 Micron (Certified), Fisher Chemical

Catalog No. t5552 Shop All Fisherbrand Products
Click to view available options
Quantity:
4.5 lb.
This item is not returnable. View return policy

Catalog No. T555-2

This item is not returnable. View return policy

Specifications

Physical Form Solid
Chemical Name or Material TissuePrep™ 2 Micron
Melting Point 53°C
Boiling Point 322°C
CAS 64742-43-4, 9017-27-0
Packaging Carton
Solubility Solubility in water: insoluble
Color White
Quantity 4.5 lb.
MDL Number MFCD00212755
Grade Certified
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.