Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium AA Standard, 1000 ppm Ti in 15% HCl, Ricca Chemical
SDP

Catalog No. p-3252583 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle

Chemical Identifiers

CAS 7440-32-6, 7647-01-0, 7732-18-5
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Ti
CAS Min % 69.0, 28.62, 0.07
CAS Max % 72.0, 29.79, 0.07
Equivalence 1 mL = 1 mg Ti
Physical Form Liquid
Color Colorless to Purple
Melting Point Approximately 0°C
Quantity 100 mL
Chemical Name or Material Titanium AA Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.