Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium boride, 99.5% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4455809

Quantity 10g

 View more versions of this product

Catalog No. AA4455809


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12045-63-5
99.5% (metals basis)
69.5
Titanium diboride
-325 Mesh Powder
B2Ti
MFCD00049595
Titanium boride

Specifications

12045-63-5
99.5% (metals basis)
69.5
Titanium diboride
Titanium boride
-325 Mesh Powder
B2Ti
MFCD00049595
10g

Safety and Handling

SDS & Safety Documents

EINECSNumber : 234-961-4

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit