Learn More
Titanium boride, Thermo Scientific Chemicals
Description
Titanium boride is used in seals wear parts and cutting tools. It is used to make ballistic armour due to its high hardness. It is used to constitute composite materials in which the presence of the material serves to increase strength and fracture toughness of the matrix.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 3225°C |
Physical Form | Powder |
Quantity | 250 g |
Solubility Information | Soluble in hydrogen peroxide and sulfuric acid. |
Formula Weight | 69.5 |
Density | 4.38 |
Chemical Name or Material | Titanium Boride |
Safety and Handling
EINECSNumber | 234-961-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.