Learn More
Titanium(II) hydride, 99% (metals basis)
Description
Titanium(II) hydride is widely utilized as an additive in the manufacturing of Alnico sintered magnets and in sintered powdered metals. It is used in the production of metal foam, powdered titanium metal and in pyrotechnics. It is involved in various applications such as ceramics, pyrotechnics and sports equipment. It is also used as a laboratory reagent and as a blowing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 450°C (decomposition) |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 99% (metals basis) |
| Solubility Information | Soluble in water. |
| Formula Weight | 49.92 |
| Sensitivity | Air and moisture sensitive |
| Density | 3.9 |
| Chemical Name or Material | Titanium(II) hydride |
Safety and Handling
| Hazard Category | H228-H500 |
| Precautionary Statement | P210-P240-P241-P280-P370+P378q |
| DOTInformation | Hazard Class: 4.1; Packaging Group: II |
| EINECSNumber | 231-726-8 |
| RTECSNumber | XR2130000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.