Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(II) oxide, 99.5% (metals basis)

Catalog No. AA7712622
Click to view available options
Quantity:
5 g
25 g
100 g
500 g

CAS: 12137-20-1 | OTi | 63.87 g/mol

Titanium(II) oxide associated with vanadium oxide forms a transition-metal-oxide monolayer catalyst, which is used in the selective oxidation and ammoxidation of alkylaromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12137-20-1
Molecular Formula OTi
Molecular Weight (g/mol) 63.87
MDL Number MFCD00036279
InChI Key OGIDPMRJRNCKJF-UHFFFAOYSA-N
Synonym titanium monoxide, titanium ii oxide, titanium oxide tio, oxido titanium, titanium ii oxide pellets, titanium oxide, 99.9 %, titanium ii oxide, optical grade, titanium ii oxide trace metals basis 5g, titanium ii oxide, coating quality, granular, titanium ii oxide, pieces trace metals basis
PubChem CID 61685
IUPAC Name oxotitanium
SMILES O=[Ti]

Specifications

Boiling Point >3000°C
Melting Point 1700°C
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.5%, metal basis
Solubility Information Soluble in concentrated sulfuric acid,concentrated hydrofluoric acid and alkali. Insoluble in water,diluted sulfuric acid,hydrochloric acid,nitric acid and organic solvents.
Formula Weight 63.9
Density 4.95
Chemical Name or Material Titanium(II) Oxide
EINECSNumber 235-236-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.