Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(III) chloride, 20% w/v solution in 2N Hydrochloric acid

Catalog No. p-3593535
Click to view available options
Quantity:
25 mL
250 mL
1 L
Packaging:
Plastic bottle

CAS: 7705-07-9 | Cl3Ti | 154.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Titanium(III) chloride
Color Purple
CAS 7705-07-9, 7647-01-0, 7732-18-5
Physical Form Liquid
Quantity 25 mL
Molecular Formula Cl3Ti
Linear Formula TiCl3
MDL Number MFCD00011266
Merck Index 15,9637
Synonym Titanous chloride
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, practically insoluble in ether
InChI Key YONPGGFAJWQGJC-UHFFFAOYSA-K
SMILES [Cl-].[Cl-].[Cl-].[Ti+3]
IUPAC Name titanium(3+) trichloride
Molecular Weight (g/mol) 154.22
Formula Weight 154.26
Percent Purity ∼20% (TiCl3) (exact strength stated on certificate of analysis)
Packaging Plastic bottle
Density 1.2000g/mL
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.