Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(III) Chloride-Aluminum(III) Chloride, TiCl3 76.0-78.5%, Alfa Aesar™

Click to view available options
Quantity:
100g
500g
12003-13-3
(TiCl3)3AlCl3
MFCD00145412
Titanium aluminum chloride,Aluminum titanium chloride
12003-13-3
(TiCl3)3AlCl3
MFCD00145412
Titanium aluminum chloride,Aluminum titanium chloride
UN Number UN3260
Solubility Information Insoluble in water.
Formula Weight 596.15
Physical Form Powder
Percent Purity TiCl3 76.0 to 78.5%
Packaging Glass bottle
Sensitivity Air and moisture sensitive
Quantity 500g
Chemical Name or Material Titanium(III) chloride-Aluminum(III) chloride

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 234-421-8

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon