Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(III) chloride-Aluminum(III) chloride, TiCl{3} 76.0-78.5%

Catalog No. aa3672922
Click to view available options
Quantity:
100 g
500 g

Used as a catalyst in the production of polypropylene. | CAS: 12003-13-3 | AlCl12Ti3 | 595.98 g/mol

Titanium(III) chloride-Aluminum(III) chloride, TiCl{3} 76.0-78.5% is used as a catalyst in the production of polypropylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12003-13-3
Molecular Formula AlCl12Ti3
Molecular Weight (g/mol) 595.98
MDL Number MFCD00145412
InChI Key KSKPGAGLAZXNEO-UHFFFAOYSA-H
IUPAC Name titanium(4+) aluminium(3+) heptachloride
SMILES Cl[Al](Cl)Cl.Cl[Ti](Cl)Cl

Specifications

Physical Form Powder
Quantity 100 g
Assay Percent Range 76.0 to 78.5%
Synonym Titanium aluminum chloride
Solubility Information Insoluble in water.
Formula Weight 596.15
Sensitivity Air and moisture sensitive
Chemical Name or Material Titanium(III) Chloride-Aluminum(III) Chloride
Hazard Category H314
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 234-421-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.