Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Titanium Indicator Solution, Reagents
Click to view available options
Quantity:
500 mL
1 L
4 L
Specifications
Specifications
| Chemical Name or Material | Titanium Indicator Solution |
| CAS | 7732-18-5, 5965-83-3, 7664-93-9 |
| Boiling Point | 100°C |
| Grade | Reagent |
| Packaging | Opaque HDPE |
| Specific Gravity | 1.13 |
| Color | Pink |
| Quantity | 4 L |
| Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction