Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) 2-ethylhexyloxide, 95%

Catalog No. AA77107A3
Click to view available options
Quantity:
100 g
500 g
2 kg

CAS: 1070-10-6 | C32H68O4Ti | 564.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070-10-6
Molecular Formula C32H68O4Ti
Molecular Weight (g/mol) 564.76
MDL Number MFCD00015264,MFCD00015264
InChI Key KTXWGMUMDPYXNN-UHFFFAOYNA-N
Synonym tyzor tot, 2-ethylhexyl orthotitanate, tetra 2-ethylhexyl titanate, tetra-2-ethylhexyl titanate, tetrakis 2-ethylhexyl titanate, titanic iv acid, tetrakis 2-ethylhexyl ester, 1-hexanol, titanium 4+ salt
PubChem CID 54603245
IUPAC Name titanium(4+) tetrakis(2-ethylhexan-1-olate)
SMILES [Ti+4].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-]

Specifications

Boiling Point 194°C (0.25mmHg)
Melting Point <-25°C
Color Yellow
Physical Form Liquid
Quantity 2 kg
Linear Formula Ti[OCH2CH(C2H5)(CH2)3CH3]4
Beilstein 3692023
Flash Point 71°C (160°F)
Formula Weight 564.79
Percent Purity 95%
Refractive Index 1.482
Sensitivity Moisture sensitive
Density 0.926 g/mL
Chemical Name or Material Titanium (IV) 2-ethylhexyloxide
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H317-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 213-969-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.